Assay Details
| Gene Symbol: FAM47C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017968225.1 | XP_017823714.1 | 1 - 2 | 1550 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM47C| Gene Symbol: FAM47C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017968225.1 | XP_017823714.1 | 1 - 2 | 1550 | 66 | |
Set Membership: |
|