Assay Details
| Gene Symbol: PCDHAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744252.3 | XP_002744298.1 | 1 | 3417 | 56 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PCDHAC1| Gene Symbol: PCDHAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744252.3 | XP_002744298.1 | 1 | 3417 | 56 | |
Set Membership: |
|