Assay Details
| Gene Symbol: IL18R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008980196.2 | XP_008978444.1 | 6 - 7 | 1347 | 75 | |
| XM_008980197.2 | XP_008978445.1 | 7 - 8 | 1355 | 75 | ||
| XM_008980198.2 | XP_008978446.1 | 6 - 7 | 802 | 75 | ||
| XM_008980199.2 | XP_008978447.1 | 6 - 7 | 1349 | 75 | ||
| XM_017964110.1 | XP_017819599.1 | 7 - 8 | 1649 | 75 | ||
More Information
Set Membership: |
|

IL18R1
Genomic Map