Assay Details
| Gene Symbol: PRG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009008133.1 | XP_009006381.1 | 3 - 4 | 471 | 74 | |
| XM_017978136.1 | XP_017833625.1 | 3 - 4 | 387 | 74 | ||
More Information
Set Membership: |
|

PRG2
Genomic Map