Assay Details
Gene Symbol: KIRREL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002760125.3 | XP_002760171.1 | 9 - 10 | 1565 | 82 | |
XM_008984666.2 | XP_008982914.1 | 9 - 10 | 1613 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
