Assay Details
Gene Symbol: FOXA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002747493.3 | XP_002747539.1 | 2 - 3 | 3676 | 107 | |
XM_008995833.2 | XP_008994081.1 | 3 - 4 | 3617 | 107 | ||
XM_017972002.1 | XP_017827491.1 | 2 - 3 | 3676 | 107 | ||
XM_017972003.1 | XP_017827492.1 | 2 - 3 | 3676 | 107 | ||
XM_017972004.1 | XP_017827493.1 | 2 - 3 | 3676 | 107 | ||
XM_017972005.1 | XP_017827494.1 | 2 - 3 | 3676 | 107 | ||
XM_017972006.1 | XP_017827495.1 | 2 - 3 | 3676 | 107 | ||
XM_017972007.1 | XP_017827496.1 | 2 - 3 | 3676 | 107 | ||
XM_017972008.1 | XP_017827497.1 | 2 - 3 | 3676 | 107 | ||
XM_017972009.1 | XP_017827498.1 | 2 - 3 | 3676 | 107 | ||
XM_017972010.1 | XP_017827499.1 | 2 - 3 | 3676 | 107 | ||
XM_017972011.1 | XP_017827500.1 | 2 - 3 | 3676 | 107 | ||
XM_017972012.1 | XP_017827501.1 | 2 - 3 | 3676 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
