Assay Details
| Gene Symbol: C2H5orf24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744168.4 | XP_002744214.1 | 2 - 3 | 689 | 72 | |
| XM_008985088.2 | XP_008983336.1 | 2 - 3 | 781 | 72 | ||
| XM_017967711.1 | XP_017823200.1 | 2 - 3 | 689 | 72 | ||
More Information
Set Membership: |
|

C2H5orf24
Genomic Map