Assay Details
| Gene Symbol: CYP4Z1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002750790.2 | XP_002750836.1 | 2 - 3 | 441 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CYP4Z1| Gene Symbol: CYP4Z1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002750790.2 | XP_002750836.1 | 2 - 3 | 441 | 69 | |
Set Membership: |
|