Assay Details
Gene Symbol: NXPE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_009006843.2 | XP_009005091.1 | 5 - 6 | 1312 | 113 | |
XM_017977550.1 | XP_017833039.1 | 6 - 7 | 1590 | 113 | ||
XM_017977551.1 | XP_017833040.1 | 4 - 5 | 5074 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
