Assay Details
| Gene Symbol: TGFB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002762160.3 | XP_002762206.1 | 1 - 2 | 1159 | 57 | |
| XM_008988065.1 | XP_008986313.1 | 1 - 2 | 1231 | 57 | ||
More Information
Set Membership: |
|

TGFB1
Genomic Map