Assay Details
| Gene Symbol: CCDC129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003733566.3 | XP_003733614.3 | 7 - 8 | 945 | 74 | |
| XM_017975216.1 | XP_017830705.1 | 7 - 8 | 682 | 74 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC129| Gene Symbol: CCDC129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003733566.3 | XP_003733614.3 | 7 - 8 | 945 | 74 | |
| XM_017975216.1 | XP_017830705.1 | 7 - 8 | 682 | 74 | ||