Assay Details
| Gene Symbol: TEDDM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017966021.1 | XP_017821510.1 | 1 | 2016 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TEDDM1| Gene Symbol: TEDDM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017966021.1 | XP_017821510.1 | 1 | 2016 | 67 | |
Set Membership: |
|