Assay Details
Gene Symbol: ZNF234 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008988174.2 | XP_008986422.1 | 4 - 5 | 411 | 64 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
