Assay Details
Gene Symbol: DHRS12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017971233.1 | XP_017826722.1 | 1 - 2 | 1699 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
