Assay Details
| Gene Symbol: C11H11orf65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017977519.1 | XP_017833008.1 | 7 - 8 | 802 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C11H11orf65| Gene Symbol: C11H11orf65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017977519.1 | XP_017833008.1 | 7 - 8 | 802 | 87 | |
Set Membership: |
|