Assay Details
Gene Symbol: LOC100414085 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002757475.2 | XP_002757521.1 | 3 - 4 | 511 | 101 | |
XM_017964121.1 | XP_017819610.1 | 3 - 4 | 515 | 101 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
