Assay Details
Gene Symbol: NKX6-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002745664.3 | XP_002745710.1 | 1 - 2 | 672 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
