Assay Details
| Gene Symbol: LOC100400601 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017965753.1 | XP_017821242.1 | 12 - 13 | 1959 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100400601| Gene Symbol: LOC100400601 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017965753.1 | XP_017821242.1 | 12 - 13 | 1959 | 65 | |
Set Membership: |
|