Assay Details
| Gene Symbol: LOC100400601 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017965753.1 | XP_017821242.1 | 26 - 27 | 4198 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100400601| Gene Symbol: LOC100400601 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017965753.1 | XP_017821242.1 | 26 - 27 | 4198 | 77 | |
Set Membership: |
|