Assay Details
| Gene Symbol: TMPRSS11B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017970070.1 | XP_017825559.1 | 7 - 8 | 728 | 104 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMPRSS11B| Gene Symbol: TMPRSS11B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017970070.1 | XP_017825559.1 | 7 - 8 | 728 | 104 | |
Set Membership: |
|