Assay Details
Gene Symbol: LOC103795884 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017977304.1 | XP_017832793.1 | 1 - 2 | 1336 | 74 | |
XM_017977305.1 | XP_017832794.1 | 1 - 2 | 1336 | 74 | ||
XM_017977306.1 | XP_017832795.1 | 1 - 2 | 1336 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
