Assay Details
Gene Symbol: NR0B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008989106.1 | XP_008987354.1 | 2 - 3 | 1188 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
