Assay Details
Gene Symbol: LOC100406494 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017976341.1 | XP_017831830.1 | 5 - 6 | 2686 | 60 | |
XM_017976342.1 | XP_017831831.1 | 5 - 6 | 2687 | 60 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
