Assay Details
Gene Symbol: NSD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002756961.2 | XP_002757007.1 | 11 - 12 | 2305 | 63 | |
XM_002756962.2 | XP_002757008.1 | 11 - 12 | 2638 | 63 | ||
XM_008979337.2 | XP_008977585.1 | 11 - 12 | 2293 | 63 | ||
XM_008979338.2 | XP_008977586.1 | 11 - 12 | 2878 | 63 | ||
XM_008979339.2 | XP_008977587.1 | 11 - 12 | 2475 | 63 | ||
XM_008979340.1 | XP_008977588.1 | 11 - 12 | 2305 | 63 | ||
XM_008979341.1 | XP_008977589.1 | 11 - 12 | 2305 | 63 | ||
XM_017963779.1 | XP_017819268.1 | 11 - 12 | 2212 | 63 | ||
XM_017963780.1 | XP_017819269.1 | 11 - 12 | 2305 | 63 | ||
XM_017963781.1 | XP_017819270.1 | 11 - 12 | 2430 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
