Assay Details
Gene Symbol: CST9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008995859.2 | XP_008994107.1 | 1 - 2 | 391 | 64 | |
XM_008995860.2 | XP_008994108.1 | 1 - 2 | 369 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
