Assay Details
| Gene Symbol: GTSF1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009003934.2 | XP_009002182.2 | 3 - 4 | 516 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GTSF1L| Gene Symbol: GTSF1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009003934.2 | XP_009002182.2 | 3 - 4 | 516 | 74 | |
Set Membership: |
|