Assay Details
| Gene Symbol: CCNQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002763394.3 | XP_002763440.1 | 1 - 2 | 234 | 80 | |
| XM_002763395.3 | XP_002763441.1 | 1 - 2 | 232 | 80 | ||
More Information
Set Membership: |
|

CCNQ
Genomic Map