Assay Details
Gene Symbol: LOC100407071 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002763462.4 | XP_002763508.3 | 4 | 1517 | 93 | |
XM_008990167.2 | XP_008988415.1 | 3 | 1530 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
