Assay Details
| Gene Symbol: LOC100412372 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002764080.2 | XP_002764126.2 | 5 | 1813 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100412372| Gene Symbol: LOC100412372 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002764080.2 | XP_002764126.2 | 5 | 1813 | 98 | |
Set Membership: |
|