Assay Details
Gene Symbol: EIF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_009006590.2 | XP_009004838.1 | 10 | 2619 | 139 | |
XM_009006591.2 | XP_009004839.1 | 10 | 2615 | 139 | ||
XM_017977353.1 | XP_017832842.1 | 11 | 2635 | 139 | ||
XM_017977354.1 | XP_017832843.1 | 11 | 2609 | 139 | ||
XM_017977355.1 | XP_017832844.1 | 11 | 3092 | 139 | ||
XM_017977356.1 | XP_017832845.1 | 9 | 2462 | 139 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
