Assay Details
Gene Symbol: B3GAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008994647.2 | XP_008992895.1 | 1 | 923 | 104 | |
XM_008994649.2 | XP_008992897.1 | 1 | 923 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
