Assay Details
| Gene Symbol: NDUFB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758084.4 | XP_002758130.1 | 6 | 1021 | 88 | |
| XM_008981254.2 | XP_008979502.1 | 5 | 915 | 88 | ||
More Information
Set Membership: |
|

NDUFB5
Genomic Map