Assay Details
| Gene Symbol: EIF1AX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008989059.2 | XP_008987307.1 | 7 | 1816 | 114 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EIF1AX| Gene Symbol: EIF1AX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008989059.2 | XP_008987307.1 | 7 | 1816 | 114 | |
Set Membership: |
|