Assay Details
Gene Symbol: MPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002747172.4 | XP_002747218.1 | 5 | 816 | 72 | |
XM_008995361.2 | XP_008993609.1 | 4 | 708 | 72 | ||
XM_008995362.2 | XP_008993610.1 | 5 | 702 | 72 | ||
XM_017971533.1 | XP_017827022.1 | 4 | 633 | 72 | ||
XM_017971534.1 | XP_017827023.1 | 6 | 969 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
