Assay Details
Gene Symbol: DNAJC17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002753533.2 | XP_002753579.1 | 11 | 919 | 82 | |
XM_017976920.1 | XP_017832409.1 | 11 | 918 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
