Assay Details
Gene Symbol: Ddx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172944.1 | NP_001166415.1 | 4 - 5 | 462 | 77 | |
GenBank mRNA | AB013609.1 | - | 4 - 5 | 462 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
