Assay Details
Gene Symbol: Nqo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172986.1 | NP_001166457.1 | 3 - 4 | 374 | 59 | |
XM_005004596.3 | XP_005004653.1 | 3 - 4 | 399 | 59 | ||
GenBank mRNA | AB055403.1 | - | 3 - 4 | 374 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
