Assay Details
| Gene Symbol: Cxcl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172938.1 | NP_001166409.1 | 1 - 2 | 90 | 95 | |
| XM_013143200.2 | XP_012998654.1 | 1 - 2 | 129 | 95 | ||
| GenBank mRNA | AF052004.1 | - | 1 - 2 | 90 | 95 | |
| U95809.1 | - | 1 - 2 | 90 | 95 | ||
| U95810.1 | - | 1 - 2 | 90 | 95 | ||
More Information
Set Membership: |
|

Cxcl1
Genomic Map