Assay Details
Gene Symbol: Cxcl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172938.1 | NP_001166409.1 | 2 - 3 | 216 | 55 | |
XM_013143200.2 | XP_012998654.1 | 2 - 3 | 255 | 55 | ||
GenBank mRNA | AF052004.1 | - | 2 - 3 | 216 | 55 | |
U95809.1 | - | 2 - 3 | 216 | 55 | ||
U95810.1 | - | 2 - 3 | 216 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
