Assay Details
Gene Symbol: Timp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173024.1 | NP_001166495.1 | 4 - 5 | 661 | 66 | |
GenBank mRNA | AF127803.1 | - | 4 - 5 | 661 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
