Assay Details
Gene Symbol: Opn1mw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172989.1 | NP_001166460.1 | 1 - 2 | 126 | 66 | |
GenBank mRNA | AF132042.1 | - | 1 - 2 | 126 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
