Assay Details
| Gene Symbol: Mip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172983.1 | NP_001166454.1 | 2 - 3 | 558 | 75 | |
| GenBank mRNA | AY485151.1 | - | 2 - 3 | 524 | 75 | |
| EU327787.1 | - | 2 - 3 | 558 | 75 | ||
More Information
Set Membership: |
|

Mip
Genomic Map