Assay Details
| Gene Symbol: Gnai3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172955.1 | NP_001166426.1 | 1 - 2 | 118 | 70 | |
| XM_005007752.3 | XP_005007809.1 | 1 - 2 | 238 | 70 | ||
| GenBank mRNA | D21234.1 | - | 1 - 2 | 118 | 70 | |
More Information
Set Membership: |
|

Gnai3
Genomic Map