Assay Details
| Gene Symbol: Atp1b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172918.2 | NP_001166389.1 | 4 - 5 | 625 | 73 | |
| GenBank mRNA | D84448.1 | - | 4 - 5 | 626 | 73 | |
More Information
Set Membership: |
|

Atp1b3
Genomic Map