Assay Details
| Gene Symbol: Lpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172978.1 | NP_001166449.1 | 4 - 5 | 808 | 60 | |
| GenBank mRNA | M15483.1 | - | 4 - 5 | 808 | 60 | |
More Information
Set Membership: |
|

Lpl
Genomic Map