Assay Details
Gene Symbol: Sftpa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173014.1 | NP_001166485.1 | 2 - 3 | 342 | 87 | |
GenBank mRNA | U40869.1 | - | 2 - 3 | 342 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
