Assay Details
Gene Symbol: Pla2g2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173000.1 | NP_001166471.1 | 4 - 5 | 349 | 81 | |
GenBank mRNA | X82631.1 | - | 4 - 5 | 349 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
