Assay Details
| Gene Symbol: LOC100135519 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172946.1 | NP_001166417.1 | 2 - 3 | 217 | 98 | |
| GenBank mRNA | X85112.1 | - | 2 - 3 | 217 | 98 | |
| X92742.1 | - | 2 - 3 | 217 | 98 | ||
More Information
Set Membership: |
|

LOC100135519
Genomic Map