Assay Details
| Gene Symbol: LOC100135519 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172946.1 | NP_001166417.1 | 3 - 4 | 402 | 68 | |
| GenBank mRNA | X85112.1 | - | 3 - 4 | 402 | 68 | |
| X92742.1 | - | 3 - 4 | 402 | 68 | ||
More Information
Set Membership: |
|

LOC100135519
Genomic Map