Assay Details
| Gene Symbol: Htr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172963.1 | NP_001166434.1 | 4 - 5 | 505 | 72 | |
| GenBank mRNA | Y13585.1 | - | 4 - 5 | 505 | 72 | |
More Information
Set Membership: |
|

Htr4
Genomic Map